Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1-Cyclobutanedicarboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. C076925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C076925G 25 g
C07695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C076925G Supplier TCI America Supplier No. C076925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5445-51-2
Molecular Formula C6H8O4
Molecular Weight (g/mol) 144.126
MDL Number MFCD00001325
InChI Key CCQPAEQGAVNNIA-UHFFFAOYSA-N
Synonym 1,1-cyclobutanedicarboxylic acid, h2cbdca, 1,1-cyclobutanedicarboxylicacid, cyclobutanedicarboxylic acid, 1,1-cyclobutane dicarboxylic acid, 1,1-cyclobutanedicarboxylate, cyclobutanedicarbonic acid, zlchem 254, pubchem14151, acmc-1am73
PubChem CID 2568
ChEBI CHEBI:35691
IUPAC Name cyclobutane-1,1-dicarboxylic acid
SMILES C1CC(C1)(C(=O)O)C(=O)O

Specifications

Melting Point 158°C
Color White
Quantity 25 g
UN Number 3261
Formula Weight 144.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,1-Cyclobutanedicarboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.