Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1-Dichloroethene, >95% min., Ultra Scientific

Catalog No. USRHH020
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-RHH-020 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-RHH-020 Supplier Agilent Technologies Supplier No. RHH020
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chemical Identifiers

CAS 75-35-4
Molecular Formula C2H2Cl2
Molecular Weight (g/mol) 96.938
InChI Key LGXVIGDEPROXKC-UHFFFAOYSA-N
PubChem CID 6366
ChEBI CHEBI:34031
IUPAC Name 1,1-dichloroethene
SMILES C=C(Cl)Cl

Specifications

CAS Max % 100.0
Assay Percent Range >95% min.
Solvent or Matrix Neat
Quantity 1 g
Chemical Name or Material 1, 1-Dichloroethene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.