accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClH2C

CAS RN 75-35-4

IUPAC Name: 1,1-dichloroethene
Synonyms: vinylidene chloride vinylidene chloride (II) VDC polyvinylidene chloride as-dichloroethylene chlorure de vinylidene vinyledene chloride ccris 622 rcra waste number U078 vinylidene chloride, monomer
Molecular Weight (g/mol): 96.94
Molecular Formula: C2H2Cl2
InChi Key: LGXVIGDEPROXKC-UHFFFAOYSA-N
SMILES: ClC(Cl)=C
Grade 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Solvent or Matrix 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)