Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1'-Diethyl-2,2'-cyanine Iodide 98.0+%, TCI America™
SDP

Catalog No. D44861G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D44861G 1 g
D4486200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D44861G Supplier TCI America Supplier No. D44861G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 977-96-8
Molecular Formula C23H23IN2
Molecular Weight (g/mol) 454.355
MDL Number MFCD00011971
InChI Key GMYRVMSXMHEDTL-UHFFFAOYSA-M
Synonym decynium 22
PubChem CID 71299759
ChEBI CHEBI:37993
IUPAC Name (2Z)-1-ethyl-2-[(1-ethylquinolin-1-ium-2-yl)methylidene]quinoline;iodide
SMILES CCN1C(=CC2=[N+](C3=CC=CC=C3C=C2)CC)C=CC4=CC=CC=C41.[I-]

Specifications

Melting Point 277°C
Quantity 1 g
UN Number 2811
Formula Weight 454.36
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1,1′-Diethyl-2,2′-cyanine Iodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.