Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1-Dimethyl-1,3-dihydrobenzo[c][1,2]oxasilole 98.0+%, TCI America™
SDP

Catalog No. D42431G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D42431G 1 g
D42435G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D42431G Supplier TCI America Supplier No. D42431G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 321903-29-1
Molecular Formula C9H12OSi
Molecular Weight (g/mol) 164.279
MDL Number MFCD20922818
InChI Key GLZOQCBCUKBUJG-UHFFFAOYSA-N
Synonym 1,3-Dihydro-1,1-dimethyl-2,1-benzoxasilole
PubChem CID 13085174
IUPAC Name 1,1-dimethyl-3H-2,1-benzoxasilole
SMILES C[Si]1(C2=CC=CC=C2CO1)C

Specifications

Color Colorless
Boiling Point 45°C
Quantity 1 g
Formula Weight 164.28
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,1-Dimethyl-1,3-dihydrobenzo[c][1,2]oxasilole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.