Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1-Dimethylguanidine Sulfate 98.0+%, TCI America™
SDP

Catalog No. D179925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D179925G 25 g
D17995G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D179925G Supplier TCI America Supplier No. D179925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 598-65-2
Molecular Formula C6H20N6O4S
Molecular Weight (g/mol) 272.324
MDL Number MFCD00013131
InChI Key QSCHFHVDZCPIKX-UHFFFAOYSA-N
Synonym 1,1-dimethylguanidine sulfate 2:1, guanidine, n,n-dimethyl-, sulfate 2:1, bis dimethylguanidine ; sulfuric acid, 1,1-dimethylguanidine hemisulfate, pubchem19198, guanidine,n,n-dimethyl, 1,1-dimethyl guanidinium sulfate, 1,1-dimethylguanidine sulfate salt, 1,1-dimethylguanidinium sulphate 2:1
PubChem CID 69024
IUPAC Name 1,1-dimethylguanidine;sulfuric acid
SMILES CN(C)C(=N)N.CN(C)C(=N)N.OS(=O)(=O)O

Specifications

Color White
Quantity 25 g
Formula Weight 272.32
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,1-Dimethylguanidine Sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.