Learn More
AdipoGen 1,1'-Dioctadecyl-3,3,3',3'-tetramethylindocarbocyanine perchlorate, 41085-99-8, MFCD00142354, 100 mg
SDP
Supplier:  AdipoGen CDXD0230M100
Encompass_Preferred
MF: C59H97ClN2O4, MW: 933.87, ≥98%, Appearance: Burgundy to purple powder or crystalline powder. Endoplasmic reticulum membrane stain. Lipophilic carbocyanine dye used primarily for optical recordings of membrane voltage and for studies of membrane fluidity. Retrograde stain for neurons; provides intense, long-lasting staining of live neurons in vivo and in vitro.
Catalog No. 50-596-227
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.