Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1-Diphenyl-2-thiourea 98.0+%, TCI America™
SDP

Catalog No. D091825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D091825G 25 g
D09185G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D091825G Supplier TCI America Supplier No. D091825G

Chemical Identifiers

CAS 3898-08-6
Molecular Formula C13H12N2S
Molecular Weight (g/mol) 228.31
MDL Number MFCD00059151
InChI Key FPZXQVCYHDMIIA-UHFFFAOYSA-N
Synonym N,N-Diphenylthiourea
PubChem CID 3034143
IUPAC Name 1,1-diphenylthiourea
SMILES C1=CC=C(C=C1)N(C2=CC=CC=C2)C(=S)N

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 228.31
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,1-Diphenyl-2-thiourea
RTECSNumber YT2100000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.