Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1'-Ferrocenedicarboxylic Acid, TCI America™
SDP

Catalog No. B09141G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B09141G 1 g
B09145G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B09141G Supplier TCI America Supplier No. B09141G

Chemical Identifiers

CAS 1293-87-4
Molecular Formula C12H10FeO4
Molecular Weight (g/mol) 274.05
MDL Number MFCD00001423
InChI Key OALUACVAKUHFGK-UHFFFAOYNA-N
Synonym Bis(carboxycyclopentadienyl) Iron, 1,1′-Dicarboxyferrocene
IUPAC Name 1,1'-Ferrocenedicarboxylic acid
SMILES OC(=O)C1C=CC=C1[Fe]C1=CC=CC1C(O)=O

Specifications

Quantity 1 g
Formula Weight 274.05
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,1′-Ferrocenedicarboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.