Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(1-Naphthylmethyl)piperazine, 97%

Catalog No. AAH5539806
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5539806 5 g
AAH5539803 1 g
AAH5539814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH5539806 Supplier Thermo Scientific Chemicals Supplier No. H5539806
Add to Cart

CAS: 40675-81-8 | C15H18N2 | 226.323 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40675-81-8
Molecular Formula C15H18N2
Molecular Weight (g/mol) 226.323
MDL Number MFCD01314185
InChI Key HGYDREHWXXUUIS-UHFFFAOYSA-N
Synonym 1-1-naphthylmethyl piperazine, 1-naphthalen-1-ylmethyl piperazine, 1-1-naphthylmethyl-piperazine, 1-naphthalen-1-ylmethyl-piperazine, 1-naphthalen-1-yl methyl piperazine, naphthylmethyl piperazine, 1-naphthylmethyl piperazine, 1-piperazin-1-yl naphthalene, 1-naphth-1-ylmethyl piperazine, 1-1-naphthalenylmethyl piperazine
PubChem CID 701891
IUPAC Name 1-(naphthalen-1-ylmethyl)piperazine
SMILES C1CN(CCN1)CC2=CC=CC3=CC=CC=C32

Specifications

Melting Point 63°C to 67°C
Color White to Brown
Quantity 5 g
UN Number UN3077
Formula Weight 226.32
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 1-(1-Naphthylmethyl)piperazine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 9; Packaging Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.