Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1'-Sulfonyldiimidazole, 98+%, Acros Organics

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
7189-69-7
C6H6N4O2S
198.21
MFCD00015893
ZLKNPIVTWNMMMH-UHFFFAOYSA-N
1,1'-sulfonyldiimidazole,1,1'-sulfonylbis 1h-imidazole,n,n'-sulfonyldiimidazole sdi,1,1'-sulphonylbis-1h-imidazole,1-1h-imidazole-1-sulfonyl-1h-imidazole,1h-imidazole, 1,1'-sulfonylbis,n,n'-sulfuryldiimidazole,1-imidazole-1-sulfonyl imidazole,1,1-sulfonyldiimidazole
81609
1-imidazol-1-ylsulfonylimidazole
C1=CN(C=N1)S(=O)(=O)N2C=CN=C2
7189-69-7
C6H6N4O2S
198.21
MFCD00015893
ZLKNPIVTWNMMMH-UHFFFAOYSA-N
1,1'-sulfonyldiimidazole,1,1'-sulfonylbis 1h-imidazole,n,n'-sulfonyldiimidazole sdi,1,1'-sulphonylbis-1h-imidazole,1-1h-imidazole-1-sulfonyl-1h-imidazole,1h-imidazole, 1,1'-sulfonylbis,n,n'-sulfuryldiimidazole,1-imidazole-1-sulfonyl imidazole,1,1-sulfonyldiimidazole
81609
1-imidazol-1-ylsulfonylimidazole
C1=CN(C=N1)S(=O)(=O)N2C=CN=C2
Color Cream
Melting Point 135.0°C to 137.0°C
Packaging Glass bottle
Quantity 5g
Infrared Spectrum Authentic
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98% min. (HPLC)
Formula Weight 198.21
Physical Form Crystalline Powder
Percent Purity 98+%
Chemical Name or Material 1, 1'-Sulfonyldiimidazole
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning

EINECSNumber : 230-554-