Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trifluoromethanesulfonic acid, 99%, extra pure

Catalog No. AC169892500
Change view
Click to view available options
Quantity:
1 mL
10 mL
50 mL
250 mL
1 L
Packaging:
Glass bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC169892500 250 mL Glass bottle
AC169890010 1 mL Glass bottle
AC169890100 10 mL Glass bottle
AC169890500 50 mL Glass bottle
AC169890011 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. AC169892500 Supplier Thermo Scientific Chemicals Supplier No. 169892500
Only null left
Add to Cart

Trifluoromethanesulfonic acid, 99%, CHF3O3S, CAS Number-1493-13-6, hotf, methanesulfonic acid, trifluoro, tfmsa, perfluoromethanesulfonic acid, tfoh, trifluoromethanesulphonic acid, triflic acid, trifluoromethane sulfonic acid, trimsylate, trifluoromethylsulfonic acid, 250mL, 03,IV,34, 162.0 deg.C | CAS: 1493-13-6 | CHF3O3S | 150.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1493-13-6
Molecular Formula CHF3O3S
Molecular Weight (g/mol) 150.07
MDL Number MFCD00007514
InChI Key ITMCEJHCFYSIIV-UHFFFAOYSA-N
Synonym triflic acid, perfluoromethanesulfonic acid, trifluoromethanesulphonic acid, tfmsa, trifluoromethane sulfonic acid, tfoh, methanesulfonic acid, trifluoro, hotf, trifluoromethylsulfonic acid, trimsylate
PubChem CID 62406
ChEBI CHEBI:48511
SMILES OS(=O)(=O)C(F)(F)F

Specifications

Melting Point -40.0°C
Color Colorless to Yellow
Density 1.6900g/mL
Boiling Point 162.0°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.3260 to 1.3300
Linear Formula CF3SO3H
Quantity 250 mL
Beilstein 03,IV,34
Fieser 04,532; 05,701; 06,617; 08,504; 09,531; 14,152; 15,41; 16,357; 17,24
Specific Gravity 1.69
Merck Index 15,9842
Solubility Information Solubility in water: soluble. Other solubilities: soluble in many polar organic solvents such as dim,sulfolane,dimethyl sulfoxide,dimethyl sulfone an
IUPAC Name trifluoromethanesulfonic acid
Viscosity 2.87 mPa.s (25°C)
Formula Weight 150.07
Percent Purity 99%
Grade Extra Pure
Physical Form Fuming Liquid
Chemical Name or Material Trifluoromethanesulfonic acid
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
May cause respiratory irritation.
May be corrosive to metals.
Precautionary Statement GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 216-087-5
Recommended Storage May slightly darken during storage

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.