Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,11-Diamino-3,6,9-trioxaundecane 97.0+%, TCI America™
SDP

Catalog No. D36641G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D36641G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D36641G Supplier TCI America Supplier No. D36641G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 929-75-9
Molecular Formula C8H20N2O3
Molecular Weight (g/mol) 192.26
MDL Number MFCD00628164
InChI Key NIQFAJBKEHPUAM-UHFFFAOYSA-N
Synonym Bis[2-(2-aminoethoxy)ethyl] Ether, 3,6,9-Trioxaundecane-1,11-diamine
PubChem CID 70250
IUPAC Name 2-{2-[2-(2-aminoethoxy)ethoxy]ethoxy}ethan-1-amine
SMILES NCCOCCOCCOCCN

Specifications

Color Yellow
Boiling Point 127°C
Quantity 1 g
Formula Weight 192.26
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1,11-Diamino-3,6,9-trioxaundecane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.