Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,12-Diaminododecane, 98+%

Catalog No. AAL0425806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0425806 5 g
AAL0425814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0425806 Supplier Thermo Scientific Chemicals Supplier No. L0425806
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 2783-17-7 | C12H28N2 | 200.37 g/mol

1,12-Diaminododecane is used for synthesis of medicinal drugs, polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,12-Diaminododecane is used for synthesis of medicinal drugs, polymers.

Solubility
Soluble in ethanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2783-17-7
Molecular Formula C12H28N2
Molecular Weight (g/mol) 200.37
MDL Number MFCD00008155
InChI Key QFTYSVGGYOXFRQ-UHFFFAOYSA-N
Synonym 1,12-diaminododecane, 1,12-dodecanediamine, dodecamethylenediamine, dodecyldiamine, 1,12-dodecamethylenediamine, 1,12-diamindodecane, 1,12'-dodecylenediamine, 1,12'-dodecamethylenediamine, unii-j3lm80w9nt, 1,12-dodecylenediamine
PubChem CID 17727
ChEBI CHEBI:49385
IUPAC Name dodecane-1,12-diamine
SMILES C(CCCCCCN)CCCCCN

Specifications

Melting Point 68°C to 72°C
Flash Point 155°C (311°F)
Linear Formula H2N(CH2)12NH2
Quantity 5 g
UN Number UN3259
Beilstein 1742765
Solubility Information Soluble in ethanol.
Formula Weight 200.37
Percent Purity ≥98%
Chemical Name or Material 1,12-Diaminododecane
Hazard Category H302-H314-H317-H335
Hazard Statement GHS H Statement
H314-H318-H302-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
EINECSNumber 220-489-6
RTECSNumber JR2200000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.