Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,12-Dodecanediol 99.0+%, TCI America™
SDP

Catalog No. D0969100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0969100G 100 g
D096925G 25 g
D0969500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D0969100G Supplier TCI America Supplier No. D0969100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5675-51-4
Molecular Formula C12H26O2
Molecular Weight (g/mol) 202.34
MDL Number MFCD00004755
InChI Key GHLKSLMMWAKNBM-UHFFFAOYSA-N
Synonym 1,12-dodecanediol, dodecamethylene glycol, chembl17440, 1,12-dihydroxy dodecane, 1,12-dodecandiol, 1,12-dodecane diol, acmc-1ambo, 1,12-dihydroxydodecane, ksc270c5h
PubChem CID 79758
IUPAC Name dodecane-1,12-diol
SMILES OCCCCCCCCCCCCO

Specifications

Melting Point 82°C
Color White
Boiling Point 189°C
Quantity 100 g
Formula Weight 202.34
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,12-Dodecanediol
EINECSNumber (2)-0240
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.