Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,14-Diamino-3,6,9,12-tetraoxatetradecane 97.0+%, TCI America™
SDP

Catalog No. T3198100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3198100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T3198100MG Supplier TCI America Supplier No. T3198100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 68960-97-4
Molecular Formula C10H24N2O4
Molecular Weight (g/mol) 236.312
InChI Key IFZOPNLVYZYSMQ-UHFFFAOYSA-N
Synonym 3,6,9,12-Tetraoxatetradecane-1,14-diamine, Triethylene Glycol Bis(2-aminoethyl) Ether
PubChem CID 10198598
IUPAC Name 2-[2-[2-[2-(2-aminoethoxy)ethoxy]ethoxy]ethoxy]ethanamine
SMILES C(COCCOCCOCCOCCN)N

Specifications

Color Colorless
Boiling Point 139°C
Quantity 100 mg
UN Number 2735
Formula Weight 236.31
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,14-Diamino-3,6,9,12-tetraoxatetradecane
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.