Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-[2-(2-Hydroxyethoxy)ethyl]piperazine 98.0+%, TCI America™
SDP

Catalog No. H119225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H119225G 25 g
H11925G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H119225G Supplier TCI America Supplier No. H119225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13349-82-1
Molecular Formula C8H18N2O2
Molecular Weight (g/mol) 174.244
MDL Number MFCD00074931
InChI Key FLNQAPQQAZVRDA-UHFFFAOYSA-N
Synonym 1-2-2-hydroxyethoxy ethyl piperazine, 2-2-piperazin-1-yl ethoxy ethanol, 2-2-piperazin-1-ylethoxy ethanol, 2-2-1-piperazinyl ethoxy ethanol, 1-2-2-hydroxyethoxyl-ethyl piperazine, 1-2-2-hydroxyethoxy-ethyl-piperazine, ethanol, 2-2-1-piperazinyl ethoxy
PubChem CID 139436
IUPAC Name 2-(2-piperazin-1-ylethoxy)ethanol
SMILES C1CN(CCN1)CCOCCO

Specifications

Color Yellow
Boiling Point 172°C
Quantity 25 g
Formula Weight 174.24
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-[2-(2-Hydroxyethoxy)ethyl]piperazine
EINECSNumber (5)-3830
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.