Learn More
Thermo Scientific Chemicals 1,2,3,4,6-Penta-O-acetyl-D-mannopyranose, 98%
Description
1,2,3,4,6-Penta-O-acetyl-D-mannopyranose is used for the preparation of glycopyranoside phosphates for the study of T-lymphocyte mediated inflammatory diseases.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1,2,3,4,6-Penta-O-acetyl-D-mannopyranose is used for the preparation of glycopyranoside phosphates for the study of T-lymphocyte mediated inflammatory diseases.
Solubility
Soluble in dichloromethane, ether, ethyl acetate, methanol, and chloroform.
Notes
Keep container tightly closed in a dry and well-ventilated place. Store in cool place. Store at -20°C.
Specifications
Specifications
| Melting Point | 66°C to 72°C |
| Color | White |
| Quantity | 25 g |
| Solubility Information | Soluble in dichloromethane,ether,ethyl acetate,methanol,and chloroform. |
| Optical Rotation | +54° (c=1 in Chloroform) |
| Formula Weight | 390.34 |
| Percent Purity | 98% |
| Physical Form | Powder |
| Chemical Name or Material | 1,2,3,4,6-Penta-O-acetyl-D-mannopyranose |
Safety and Handling
| TSCA | No |
| Recommended Storage | Keep cold |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.