Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,3,4-Tetrahydrocarbazole 99.0+%, TCI America™
SDP

Catalog No. T1006100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1006100G 100 g
T100625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T1006100G Supplier TCI America Supplier No. T1006100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 942-01-8
Molecular Formula C12H13N
Molecular Weight (g/mol) 171.243
MDL Number MFCD00004959
InChI Key XKLNOVWDVMWTOB-UHFFFAOYSA-N
Synonym 1,2,3,4-tetrahydrocarbazole, 2,3-tetramethyleneindole, 2,3-tetramethylene-1h-indole, 5,6,7,8-tetrahydrocarbazole, tetrahydrocarbazole, 1h-carbazole, 2,3,4,9-tetrahydro, carbazole, 1,2,3,4-tetrahydro, 1h-indole, 2,3-1,4-butanediyl, carbazole, 5,6,7,8-tetrahydro, unii-8zlk0tsx93
PubChem CID 13664
IUPAC Name 2,3,4,9-tetrahydro-1H-carbazole
SMILES C1CCC2=C(C1)C3=CC=CC=C3N2

Specifications

Melting Point 119°C
Color White-Yellow
Boiling Point 186°C
Quantity 100 g
Formula Weight 171.24
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2,3,4-Tetrahydrocarbazole
RTECSNumber FE6300000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.