Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,3,4-Tetrahydroquinaldine 97.0+%, TCI America™
SDP

Catalog No. T011225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T011225G 25 g
T0112500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T011225G Supplier TCI America Supplier No. T011225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1780-19-4
Molecular Formula C10H13N
Molecular Weight (g/mol) 147.22
MDL Number MFCD00023886
InChI Key JZICUKPOZUKZLL-UHFFFAOYNA-N
Synonym 2-Methyl-1,2,3,4-tetrahydroquinoline, 1,2,3,4-Tetrahydro-2-methylquinoline
PubChem CID 96289
IUPAC Name 2-methyl-1,2,3,4-tetrahydroquinoline
SMILES CC1CCC2=CC=CC=C2N1

Specifications

Color Yellow
Boiling Point 250°C
Quantity 25 g
Formula Weight 147.22
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2,3,4-Tetrahydroquinaldine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.