Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,3-Cyclohexanetriol (cis- and trans- mixture) 97.0+%, TCI America™
SDP

Catalog No. C113210G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C113210G 10 g
C113225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C113210G Supplier TCI America Supplier No. C113210G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6286-43-7
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.159
MDL Number MFCD00060111
InChI Key IZSANPWSFUSNMY-UHFFFAOYSA-N
Synonym Hexahydropyrogallol, Pyrogallitol
PubChem CID 222962
IUPAC Name cyclohexane-1,2,3-triol
SMILES C1CC(C(C(C1)O)O)O

Specifications

Quantity 10 g
Formula Weight 132.16
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2,3-Cyclohexanetriol (cis- and trans- mixture)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.