Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2,3-Dichlorophenyl)piperazine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. D36605G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D36605G 5 g
D366025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D36605G Supplier TCI America Supplier No. D36605G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 119532-26-2
Molecular Formula C10H13Cl3N2
Molecular Weight (g/mol) 267.578
MDL Number MFCD00190238
InChI Key CYQFNNSFAGXCEC-UHFFFAOYSA-N
Synonym 1-2,3-dichlorophenyl piperazine hydrochloride, 1-2,3-dichlorophenyl piperazine monohydrochloride, 1-2,3-dichlorophenyl-piperazine hydrochloride, 2,3-dichlorophenylpiperazine hydrochloride, 1-2,3-dichlorophenyl-piperazine monohydrochloride, 1-2,3-dichlorophenyl piperazinehydrochloride, piperazine, 1-2,3-dichlorophenyl-, monohydrochloride, 1-2,3-dichlorophenyl piperazine;hydrochloride, 2,3-dichlorophenyl piperazine, chloride, zlchem 392
PubChem CID 11630372
IUPAC Name 1-(2,3-dichlorophenyl)piperazine;hydrochloride
SMILES C1CN(CCN1)C2=C(C(=CC=C2)Cl)Cl.Cl

Specifications

Melting Point 247°C
Color White-Yellow
Quantity 5 g
Formula Weight 267.58
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1-(2,3-Dichlorophenyl)piperazine Hydrochloride
EINECSNumber (5)-6790
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.