Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,2,3-Trimethylbenzene

Catalog No. T7320250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T7320250ML 50 mL
T732025ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T7320250ML Supplier Sigma Aldrich Supplier No. T7320250ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,2,3-Trimethylbenzene

Specifications

CAS 526-73-8
Melting Point -25°C (lit.)
Density 0.894 g/mL (at 25°C (literature))
Boiling Point 175°C to 176°C (lit.)
Molecular Formula C9H12
Refractive Index n20/D 1.513 (literature)
Linear Formula C6H3(CH3)3
MDL Number MFCD00008520
Quantity 50 mL
Synonym Hemellitol
Molecular Weight (g/mol) 120.19
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.