Learn More
Cayman Chemical 1 2 3TrI10 ZHeptadecenoyl 10mg
SDP
Supplier:  Cayman Chemical 2699610 MG
Encompass_Preferred
A triacylglycerol; has been used as an internal standard for the quantification of triglycerides in green microalgae and phospholipid bilayers
Catalog No. 50-293-7122