Learn More
Cayman Chemical 1 2 3TrIpalmItold31 GlcR 10mg
SDP
Supplier:  Cayman Chemical 2859310 MG
Encompass_Preferred
An internal standard for the quantification of 1,2,3-tripalmitoyl glycerol by GC- or LC-MS
Catalog No. 50-293-8524