Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,4,5-Tetracyanobenzene 98.0+%, TCI America™
SDP

Catalog No. T09881G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T09881G 1 g
T09885G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T09881G Supplier TCI America Supplier No. T09881G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 712-74-3
Molecular Formula C10H2N4
Molecular Weight (g/mol) 178.154
MDL Number MFCD00043481
InChI Key FAAXSAZENACQBT-UHFFFAOYSA-N
Synonym 1,2,4,5-tetracyanobenzene, 1,2,4,5-benzenetetracarbonitrile, tetracyanobenzene, pyromellitic nitrile, pyromellitotetranitrile, pyromellitic tetranitrile, pyromellitic acid tetranitrile, 1,2,4,5-benzenetetranitrile, pyromellitonitrile, s-tetracyanobenzene
PubChem CID 12838
IUPAC Name benzene-1,2,4,5-tetracarbonitrile
SMILES C1=C(C(=CC(=C1C#N)C#N)C#N)C#N

Specifications

Melting Point 265°C
Color White-Yellow
Quantity 1 g
UN Number 3439
Formula Weight 178.15
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1,2,4,5-Tetracyanobenzene
RTECSNumber DB9100000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.