Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,4,6-Tetra-O-acetyl-3-O-allyl-beta-D-glucopyranose 98.0+%, TCI America™
SDP

Catalog No. T24491G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T24491G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T24491G Supplier TCI America Supplier No. T24491G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 39698-00-5
Molecular Formula C17H24O10
Molecular Weight (g/mol) 388.37
MDL Number MFCD11112188
InChI Key OIMJIPHVXXKVLL-UHFFFAOYNA-N
PubChem CID 11132857
IUPAC Name [3,5,6-tris(acetyloxy)-4-(prop-2-en-1-yloxy)oxan-2-yl]methyl acetate
SMILES CC(=O)OCC1OC(OC(C)=O)C(OC(C)=O)C(OCC=C)C1OC(C)=O

Specifications

Melting Point 119°C
Color White
Quantity 1 g
Formula Weight 388.37
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2,4,6-Tetra-O-acetyl-3-O-allyl-beta-D-glucopyranose
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.