Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2,4,6-Triisopropylbenzenesulfonyl)imidazole 98.0+%, TCI America™
SDP

Catalog No. T141025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T141025G 25 g
T14105G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T141025G Supplier TCI America Supplier No. T141025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 50257-40-4
Molecular Formula C18H26N2O2S
Molecular Weight (g/mol) 334.478
MDL Number MFCD00005283
InChI Key AGGRGODMKWLSDE-UHFFFAOYSA-N
Synonym 1-2,4,6-triisopropylphenyl sulfonyl-1h-imidazole, 1-2,4,6-triisopropylbenzenesulfonyl imidazole, 1-2,4,6-triisopropylphenylsulfonyl imidazole, tpsi, 1-2',4',6'-triisopropylbenzenesulphonyl imidazole, 1-2,4,6-tri propan-2-yl phenyl sulfonylimidazole, tpsi 1-2,4,6-triisopropylbenzenesulfonyl imidazole, 1h-imidazole, 1-2,4,6-tris 1-methylethyl phenyl sulfonyl, 1-2,4,6-trisopropylbenzenesulfonyl imidazole
PubChem CID 521274
IUPAC Name 1-[2,4,6-tri(propan-2-yl)phenyl]sulfonylimidazole
SMILES CC(C)C1=CC(=C(C(=C1)C(C)C)S(=O)(=O)N2C=CN=C2)C(C)C

Specifications

Melting Point 119°C
Color White
Quantity 25 g
Formula Weight 334.48
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(2,4,6-Triisopropylbenzenesulfonyl)imidazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.