Learn More
Sigma Organic Chemistry 1 2 4-Oxadiazole-3-carb | 1G | 39512-80-6 | MFCD00457756
Supplier: Sigma Organic Chemistry CBR004211G

1 2 4-Oxadiazole-3-carb , 1G
About this Item:
CAS #: 39512-80-6
MDL #: MFCD00457756
Molecular Weight: 129.14
UNSPSC Code: 12352200
Chemical Formula: C3H3N3OS
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.