Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,4-Trichlorobenzene, 99%

Catalog No. p-7052390
Change view
Click to view available options
Quantity:
500 g
2500 g
4 x 2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1013036 500 g
AAA101300E 2500 g
AAA10130CLL 4 x 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1013036 Supplier Thermo Scientific Chemicals Supplier No. A1013036
Only null left
Add to Cart

CAS: 120-82-1 | C6H3Cl3 | 181.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
1,2,4-Trichlorobenzene is used as a dielectric and heat transfer fluid in transformers. It acts as an intermediate, degreaser, wood preservative and solvent for dye. It is a high-temperature solvent used in gel permeation chromatography, especially for polyethylene and polypropylene. Further, it is used as a lubricant and as a synthetic transformer oil.

Solubility
Miscible with ether, benzene, CS{2}.Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-82-1
Molecular Formula C6H3Cl3
Molecular Weight (g/mol) 181.44
MDL Number MFCD00000547
InChI Key PBKONEOXTCPAFI-UHFFFAOYSA-N
Synonym benzene, 1,2,4-trichloro, unsym-trichlorobenzene, hostetex l-pec, trojchlorobenzen, 1,2,4-trichlorbenzol, 1,2,4-trichlorobenzol, 1,3,4-trichlorobenzene, trichlorobenzene a, 1,2,5-trichlorobenzene, as-trichlorobenzene
PubChem CID 13
ChEBI CHEBI:28222
IUPAC Name 1,2,4-trichlorobenzene
SMILES C1=CC(=C(C=C1Cl)Cl)Cl

Specifications

Melting Point 17°C
Density 1.454
Quantity 500 g
Physical Form Liquid
Refractive Index 1.572
UN Number UN2321
Merck Index 14,9631
Boiling Point 213°C
Solubility Information Miscible with ether,benzene,CS2;Immiscible with water.
Formula Weight 181.45
Percent Purity 99%
Chemical Name or Material 1,2,4-Trichlorobenzene
Show More Show Less
Hazard Category H302-H315-H319
Hazard Statement GHS H Statement
H302-H315
Harmful if swallowed.
Causes skin irritation.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TRICHLOROBENZENES, LIQUID
EINECSNumber 204-428-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.