CAS RN 120-82-1
1,2,4-Trichlorobenzene (HPLC), Fisher Chemical
CAS: 120-82-1 Molecular Formula: C6H3Cl3 Molecular Weight (g/mol): 181.44 MDL Number: MFCD00000547 InChI Key: PBKONEOXTCPAFI-UHFFFAOYSA-N Synonym: benzene, 1,2,4-trichloro,unsym-trichlorobenzene,hostetex l-pec,trojchlorobenzen,1,2,4-trichlorbenzol,1,2,4-trichlorobenzol,1,3,4-trichlorobenzene,trichlorobenzene a,1,2,5-trichlorobenzene,as-trichlorobenzene PubChem CID: 13 ChEBI: CHEBI:28222 IUPAC Name: 1,2,4-trichlorobenzene SMILES: C1=CC(=C(C=C1Cl)Cl)Cl
1,2,4-Trichlorobenzene, 99%
CAS: 120-82-1 Molecular Formula: C6H3Cl3 Molecular Weight (g/mol): 181.44 MDL Number: MFCD00000547 InChI Key: PBKONEOXTCPAFI-UHFFFAOYSA-N Synonym: benzene, 1,2,4-trichloro,unsym-trichlorobenzene,hostetex l-pec,trojchlorobenzen,1,2,4-trichlorbenzol,1,2,4-trichlorobenzol,1,3,4-trichlorobenzene,trichlorobenzene a,1,2,5-trichlorobenzene,as-trichlorobenzene PubChem CID: 13 ChEBI: CHEBI:28222 IUPAC Name: 1,2,4-trichlorobenzene SMILES: C1=CC(=C(C=C1Cl)Cl)Cl
1,2,4-Trichlorobenzene, Spectrophotometric Grade, 99% min
CAS: 120-82-1 Molecular Formula: C6H3Cl3 Molecular Weight (g/mol): 181.44 MDL Number: MFCD00000547 InChI Key: PBKONEOXTCPAFI-UHFFFAOYSA-N Synonym: benzene, 1,2,4-trichloro,unsym-trichlorobenzene,hostetex l-pec,trojchlorobenzen,1,2,4-trichlorbenzol,1,2,4-trichlorobenzol,1,3,4-trichlorobenzene,trichlorobenzene a,1,2,5-trichlorobenzene,as-trichlorobenzene PubChem CID: 13 ChEBI: CHEBI:28222 IUPAC Name: 1,2,4-trichlorobenzene SMILES: C1=CC(=C(C=C1Cl)Cl)Cl
1,2,4-Trichlorobenzene, OmniSolv™ MilliporeSigma™
CAS: 120-82-1 Molecular Formula: C6H3Cl3 Molecular Weight (g/mol): 181.44 InChI Key: PBKONEOXTCPAFI-UHFFFAOYSA-N Synonym: benzene, 1,2,4-trichloro,unsym-trichlorobenzene,hostetex l-pec,trojchlorobenzen,1,2,4-trichlorbenzol,1,2,4-trichlorobenzol,1,3,4-trichlorobenzene,trichlorobenzene a,1,2,5-trichlorobenzene,as-trichlorobenzene PubChem CID: 13 ChEBI: CHEBI:28222 IUPAC Name: 1,2,4-trichlorobenzene SMILES: C1=CC(=C(C=C1Cl)Cl)Cl
1,2,4-Trichlorobenzene, 99%, pure
CAS: 120-82-1 MDL Number: MFCD00000547 InChI Key: PBKONEOXTCPAFI-UHFFFAOYSA-N Synonym: benzene, 1,2,4-trichloro,unsym-trichlorobenzene,hostetex l-pec,trojchlorobenzen,1,2,4-trichlorbenzol,1,2,4-trichlorobenzol,1,3,4-trichlorobenzene,trichlorobenzene a,1,2,5-trichlorobenzene,as-trichlorobenzene PubChem CID: 13 ChEBI: CHEBI:28222 IUPAC Name: 1,2,4-trichlorobenzene SMILES: C1=CC(=C(C=C1Cl)Cl)Cl
1,2,4-Trichlorobenzene, ReagentPlus™, ≥99%, Solstice
CAS: 120-82-1 Molecular Formula: C6H3Cl3 Molecular Weight (g/mol): 181.44 MDL Number: MFCD00000547 InChI Key: PBKONEOXTCPAFI-UHFFFAOYSA-N Synonym: benzene, 1,2,4-trichloro,unsym-trichlorobenzene,hostetex l-pec,trojchlorobenzen,1,2,4-trichlorbenzol,1,2,4-trichlorobenzol,1,3,4-trichlorobenzene,trichlorobenzene a,1,2,5-trichlorobenzene,as-trichlorobenzene PubChem CID: 13 ChEBI: CHEBI:28222 IUPAC Name: 1,2,4-trichlorobenzene SMILES: C1=CC(=C(C=C1Cl)Cl)Cl
1,2,4-Trichlorobenzene, 99.0% min., ACS, MilliporeSigma™
CAS: 120-82-1 Molecular Formula: C6H3Cl3 Molecular Weight (g/mol): 181.44 InChI Key: PBKONEOXTCPAFI-UHFFFAOYSA-N Synonym: benzene, 1,2,4-trichloro,unsym-trichlorobenzene,hostetex l-pec,trojchlorobenzen,1,2,4-trichlorbenzol,1,2,4-trichlorobenzol,1,3,4-trichlorobenzene,trichlorobenzene a,1,2,5-trichlorobenzene,as-trichlorobenzene PubChem CID: 13 ChEBI: CHEBI:28222 IUPAC Name: 1,2,4-trichlorobenzene SMILES: C1=CC(=C(C=C1Cl)Cl)Cl
VOCs - Sandy Loam, MilliporeSigma™ Supelco™
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
1,2,4-Trichlorobenzene, Packaged under Argon, HPLC Grade, J.T. Baker™
CAS: 120-82-1 Molecular Formula: C6H3Cl3 Molecular Weight (g/mol): 181.44 InChI Key: PBKONEOXTCPAFI-UHFFFAOYSA-N Synonym: benzene, 1,2,4-trichloro,unsym-trichlorobenzene,hostetex l-pec,trojchlorobenzen,1,2,4-trichlorbenzol,1,2,4-trichlorobenzol,1,3,4-trichlorobenzene,trichlorobenzene a,1,2,5-trichlorobenzene,as-trichlorobenzene PubChem CID: 13 ChEBI: CHEBI:28222 IUPAC Name: 1,2,4-trichlorobenzene SMILES: C1=CC(=C(C=C1Cl)Cl)Cl
1,2,4-Trichlorobenzene, for GPC, GC and spectrophotometry, >98.0%, Solstice
CAS: 120-82-1 Molecular Formula: C6H3Cl3 Molecular Weight (g/mol): 181.44 InChI Key: PBKONEOXTCPAFI-UHFFFAOYSA-N Synonym: benzene, 1,2,4-trichloro,unsym-trichlorobenzene,hostetex l-pec,trojchlorobenzen,1,2,4-trichlorbenzol,1,2,4-trichlorobenzol,1,3,4-trichlorobenzene,trichlorobenzene a,1,2,5-trichlorobenzene,as-trichlorobenzene PubChem CID: 13 ChEBI: CHEBI:28222 IUPAC Name: 1,2,4-trichlorobenzene SMILES: C1=CC(=C(C=C1Cl)Cl)Cl
1,2,4-Trichlorobenzene 98.0+%, TCI America™
CAS: 120-82-1 Molecular Formula: C6H3Cl3 Molecular Weight (g/mol): 181.44 MDL Number: MFCD00000547 InChI Key: PBKONEOXTCPAFI-UHFFFAOYSA-N Synonym: benzene, 1,2,4-trichloro,unsym-trichlorobenzene,hostetex l-pec,trojchlorobenzen,1,2,4-trichlorbenzol,1,2,4-trichlorobenzol,1,3,4-trichlorobenzene,trichlorobenzene a,1,2,5-trichlorobenzene,as-trichlorobenzene PubChem CID: 13 ChEBI: CHEBI:28222 IUPAC Name: 1,2,4-trichlorobenzene SMILES: C1=CC(=C(C=C1Cl)Cl)Cl
Base-Neutrals Matrix Spike Tinted Indicator, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
1,2,4-Trichlorobenzene, CHROMASOLV™, ≥99%, Solstice
CAS: 120-82-1 Molecular Formula: C6H3Cl3 Molecular Weight (g/mol): 181.44 MDL Number: MFCD00000547 InChI Key: PBKONEOXTCPAFI-UHFFFAOYSA-N Synonym: benzene, 1,2,4-trichloro,unsym-trichlorobenzene,hostetex l-pec,trojchlorobenzen,1,2,4-trichlorbenzol,1,2,4-trichlorobenzol,1,3,4-trichlorobenzene,trichlorobenzene a,1,2,5-trichlorobenzene,as-trichlorobenzene PubChem CID: 13 ChEBI: CHEBI:28222 IUPAC Name: 1,2,4-trichlorobenzene SMILES: C1=CC(=C(C=C1Cl)Cl)Cl