Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,4-Trichlorobenzene, CHROMASOLV™, ≥99%, Solstice

Catalog No. 60045708 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-708 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-045-708 Supplier Solstice Supplier No. 2564121L

missing translation for 'validMainframeMsg'

CHROMASOLV™,≥99%

  • High purity chromatography solvents
  • Suitability tested for specified use from HPLC up to UHPLC-MS applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-82-1
Molecular Formula C6H3Cl3
Molecular Weight (g/mol) 181.44
MDL Number MFCD00000547
InChI Key PBKONEOXTCPAFI-UHFFFAOYSA-N
Synonym benzene, 1,2,4-trichloro, unsym-trichlorobenzene, hostetex l-pec, trojchlorobenzen, 1,2,4-trichlorbenzol, 1,2,4-trichlorobenzol, 1,3,4-trichlorobenzene, trichlorobenzene a, 1,2,5-trichlorobenzene, as-trichlorobenzene
PubChem CID 13
ChEBI CHEBI:28222
IUPAC Name 1,2,4-trichlorobenzene
SMILES C1=CC(=C(C=C1Cl)Cl)Cl

Specifications

Melting Point 16°C (literature)
CAS Max % ≤100.0000%
Density 1.454 g/mL at 25 °C(lit.)
Quantity 1 L
Flash Point 105°C
Packaging Glass Bottle
Linear Formula C6H3Cl3
UN Number UN2321
Beilstein 956819
Boiling Point 213.5°C
Formula Weight 181.46
Percent Purity >99%
Grade HPLC
Chemical Name or Material 1,2,4-Trichlorobenzene, CHROMASOLV™, ≥99%
Show More Show Less
ShelfLife 1620 days from date of manufacture
DOTInformation Transport Hazard Class: 6.1; Packing Group:3: III; Proper Shipping Name: Trichlorobenzenes, liquid
EINECSNumber 204-428-0
Recommended Storage Room Temp
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.