Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,4-Trichlorobenzene, Packaged under Argon, HPLC Grade, J.T. Baker™

Catalog No. 02002006 Shop All Avantor Products
Change view
Click to view available options
Quantity:
3.8 L
Packaging:
Narrow Mouth PVC Coated Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-002-006 3.8 L Narrow Mouth PVC Coated Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-002-006 Supplier Avantor Supplier No. 944405
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High purity solvents for use in analytical and preparative separations

    Suitable for use in gel permeation chromatography

Chemical Identifiers

CAS 120-82-1
Molecular Formula C6H3Cl3
Molecular Weight (g/mol) 181.44
InChI Key PBKONEOXTCPAFI-UHFFFAOYSA-N
Synonym benzene, 1,2,4-trichloro, unsym-trichlorobenzene, hostetex l-pec, trojchlorobenzen, 1,2,4-trichlorbenzol, 1,2,4-trichlorobenzol, 1,3,4-trichlorobenzene, trichlorobenzene a, 1,2,5-trichlorobenzene, as-trichlorobenzene
PubChem CID 13
ChEBI CHEBI:28222
IUPAC Name 1,2,4-trichlorobenzene
SMILES C1=CC(=C(C=C1Cl)Cl)Cl

Specifications

Quantity 3.8 L
Packaging Narrow Mouth PVC Coated Amber Glass Bottle
Specific Gravity 1L = 1.45kg
Formula Weight 181.45
Grade HPLC
Chemical Name or Material 1,2,4-Trichlorobenzene, Packaged under Argon
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.