Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,4-Trichlorobenzene, ReagentPlus™, ≥99%, Solstice

Catalog No. 60044786 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
2.5 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-044-786 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-044-786 Supplier Solstice Supplier No. 1320472.5L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Reagent Grade,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-82-1
Molecular Formula C6H3Cl3
Molecular Weight (g/mol) 181.44
MDL Number MFCD00000547
InChI Key PBKONEOXTCPAFI-UHFFFAOYSA-N
Synonym benzene, 1,2,4-trichloro, unsym-trichlorobenzene, hostetex l-pec, trojchlorobenzen, 1,2,4-trichlorbenzol, 1,2,4-trichlorobenzol, 1,3,4-trichlorobenzene, trichlorobenzene a, 1,2,5-trichlorobenzene, as-trichlorobenzene
PubChem CID 13
ChEBI CHEBI:28222
IUPAC Name 1,2,4-trichlorobenzene
SMILES C1=CC(=C(C=C1Cl)Cl)Cl

Specifications

Melting Point 17°C
Color Colorless
Density 1.454g/mL at 25°C
Quantity 2.5 L
Explosion Limit 6.6% (150°F)
Flash Point 110°C (230°F)
Ignition Point 1060°F
Packaging Glass Bottle
Refractive Index n20/D 1.571(lit.)
Vapor Pressure 1mmHg (40°C)
UN Number UN2321
Beilstein 956819
Boiling Point 213.5°C
Formula Weight 181.45g/mol
Percent Purity ≥99%
Grade ReagentPlus
Chemical Name or Material 1,2,4-Trichlorobenzene
Show More Show Less
Hazard Statement P301 + P312 + P330
Precautionary Statement H302-H315-H410
DOTInformation 6.1/PGIII
EINECSNumber 204-428-0
RTECSNumber DC2100000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.