Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,4-Trifluorobenzene 98.0+%, TCI America™
SDP

Catalog No. T103325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T103325G 25 g
T10335G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T103325G Supplier TCI America Supplier No. T103325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 367-23-7
Molecular Formula C6H3F3
Molecular Weight (g/mol) 132.09
MDL Number MFCD00000305
InChI Key PEBWOGPSYUIOBP-UHFFFAOYSA-N
Synonym benzene, 1,2,4-trifluoro, unii-qww4hnq0l1, 1,3,4-trifluorobenzene, qww4hnq0l1, pubchem1068, 1,4-trifluorobenzene, benzene,2,4-trifluoro, 1,2,4-trifluoro-benzene, 1,2,4-trifluorobenzene, acmc-209io5
PubChem CID 67773
IUPAC Name 1,2,4-trifluorobenzene
SMILES FC1=CC=C(F)C(F)=C1

Specifications

Color Yellow
Boiling Point 90°C
Quantity 25 g
UN Number 1993
Formula Weight 132.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2,4-Trifluorobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.