Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,4-Trimethylbenzene, MilliporeSigma™ Supelco™

Catalog No. 111020489 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1000 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0489 1000 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0489 Supplier MilliporeSigma Supelco Supplier No. 47324

Sigma-Aldrich offers 1,2,4-trimethylbenzene for a variety of applications that include, but are not limited to, environmental, food and beverage, forensics, petrochemical and pharmaceutical analyses. Purity of the chemical is about 99% unless otherwise stated.

Featured Industries: Environmental
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
Ignition Point 959°F
CAS 95-63-6
Vapor Pressure 4.5 mmHg (37.7°C)
Vapor Density 4.1 (vs. air)
Density 0.876 g/mL (at 20°C)
Flash Point 48°C
Physical Form Neat
Linear Formula C6H3(CH3)3
MDL Number MFCD00008527
Melting Point -44°C
Boiling Point 168°C
Synonym Pseudocumene
Type Organic Pollutant Standard
Quantity 1000 mg
Formula Weight 120.19
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.