Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2:5,6-Di-O-isopropylidene-alpha-D-allo-furanose, 98%, Thermo Scientific Chemicals

Catalog No. AC270960100
Change view
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC270960100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC270960100 Supplier Thermo Scientific Chemicals Supplier No. 270960100
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2595-05-3 | Molecular Formula: C12H20O6 | Molecular Weight: 260.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2595-05-3
Molecular Formula C12H20O6
Molecular Weight (g/mol) 260.28
MDL Number MFCD00135634
InChI Key KEJGAYKWRDILTF-VVULQXIFSA-N
Synonym 1,2:5,6-di-o-isopropylidene-alpha-d-allofuranose, 3ar,5s,6r,6ar-5-r-2,2-dimethyl-1,3-dioxolan-4-yl-2,2-dimethyltetrahydrofuro 2,3-d 1,3 dioxol-6-ol, 1,2,5,6-di-o-isopropylidene-alpha-d-allofuranose, 3ar,5s,6r,6ar-5-4r-2,2-dimethyl-1,3-dioxolan-4-yl-2,2-dimethyl-tetrahydrofuro 2,3-d 1,3 dioxol-6-ol, 1,2:5,6-bis-o-1-methylethylidene, 5,6-di-o-isopropylidene-alpha-d-allofuranose, 1,2:5,6-di-o-isopropylidene-?-d-allofuranose, 1,2:5,6-di-o-isopropylidene-alpha-d-ribo-hexofuranose, 1-o,2-o:5-o,6-o-diisopropylidene-alpha-d-allofuranose
PubChem CID 7157054
IUPAC Name (3aR,5S,6R,6aR)-5-[(4R)-2,2-dimethyl-1,3-dioxolan-4-yl]-2,2-dimethyl-3a,5,6,6a-tetrahydrofuro[2,3-d][1,3]dioxol-6-ol
SMILES CC1(OCC(O1)C2C(C3C(O2)OC(O3)(C)C)O)C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 73°C to 77°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 10 g
Specific Rotation Condition +37.50 (20.00°C c=1,chloroform)
Specific Rotation + 37.5
Formula Weight 260.28
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1, 2:5, 6-Di-O-isopropylidene-α-D-allo-furanose
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.