Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,5,6-Tetrabromohexane, 97%, Thermo Scientific™

Catalog No. AAB2059806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2059806 5 g
AAB2059814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2059806 Supplier Thermo Scientific Chemicals Supplier No. B2059806

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58443-86-0
Molecular Formula C6H10Br4
Molecular Weight (g/mol) 401.762
MDL Number MFCD00049137
InChI Key WPBWUVCMCYXPFI-UHFFFAOYSA-N
Synonym hexane, 1,2,5,6-tetrabromo, acmc-20ap74, hexane,1,2,5,6-tetrabromo
PubChem CID 94047
IUPAC Name 1,2,5,6-tetrabromohexane
SMILES C(CC(CBr)Br)C(CBr)Br

Specifications

Quantity 5 g
Formula Weight 401.76
Percent Purity 97%
Chemical Name or Material 1,2,5,6-Tetrabromohexane
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.