Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2,5-Dichloro-4-sulfophenyl)-3-methyl-5-pyrazolone Monohydrate 98.0+%, TCI America™
SDP

Catalog No. M066325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M066325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M066325G Supplier TCI America Supplier No. M066325G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 84-57-1
Molecular Formula C10H8Cl2N2O4S
Molecular Weight (g/mol) 323.144
MDL Number MFCD00035706
InChI Key KCEIVWKDBLAQKL-UHFFFAOYSA-N
PubChem CID 66536
IUPAC Name 2,5-dichloro-4-(3-methyl-5-oxo-4H-pyrazol-1-yl)benzenesulfonic acid
SMILES CC1=NN(C(=O)C1)C2=CC(=C(C=C2Cl)S(=O)(=O)O)Cl

Specifications

Color Yellow
Quantity 25 g
Formula Weight 323.14
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(2,5-Dichloro-4-sulfophenyl)-3-methyl-5-pyrazolone Monohydrate
EINECSNumber (5)-0352
RTECSNumber DB6040000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.