Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,2-Bis(2-chloroethoxy)ethane

Catalog No. 241628100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-162-8100G 100 g
24-162-8500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 24-162-8100G Supplier Sigma Aldrich Supplier No. 241628100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,2-bis(2-chloroethoxy)ethane was used in the synthesis of 1,1α-[1,2-ethanediylbis(oxy-1,2-ethanediyl)]bis[3-methyl-1H-imidazolium-1-yl] chloride.

Specifications

CAS 112-26-5
Density 1.197 g/mL (at 25°C (literature))
Boiling Point 235°C (lit.)
Molecular Formula C6H12Cl2O2
Refractive Index n20/D 1.461 (literature)
Linear Formula ClCH2CH2OCH2CH2OCH2CH2Cl
MDL Number MFCD00000976
Quantity 100 g
Synonym Dichlorotriethylene dioxide; Tri(ethylene glycol) dichloride; Triglycol dichloride’
Molecular Weight (g/mol) 187.06
Percent Purity 97%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.