Learn More
Strem, An Ascensus Company CAS# 528565-79-9. 500mg. (-)-1,2-Bis((2R,5R)-2,5-diphenylphospholano)ethane, min. 95% (R,R)-Ph-BPE. MFCD07369027
SDP
Supplier:  Strem, An Ascensus Company 150473500MG
Encompass
CAS# 528565-79-9. 500mg. (-)-1,2-Bis((2R,5R)-2,5-diphenylphospholano)ethane, min. 95% (R,R)-Ph-BPE. MFCD07369027. Molecular Weight: 506.60. Molecular Formula: (C16H16)PCH2CH2P(C16H16). Color/form: white solid Strem# 15-0473. http://www.strem.com/catalog/v/15-0473/
Catalog No. 50-901-09904