Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Bis(diphenylphosphino)benzene, 98%

Catalog No. AAB2110601
Change view
Click to view available options
Quantity:
0.25 g
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2110601 0.25 g
AAB2110603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2110601 Supplier Thermo Scientific Chemicals Supplier No. B2110601
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 13991-08-7 | C30H24P2 | 446.47 g/mol

It acts as a ligand for Baeyer-Villiger Oxidation, cycloadditions and reductions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It acts as a ligand for Baeyer-Villiger Oxidation, cycloadditions and reductions.

Solubility
Insoluble in water.

Notes
Air sensitive. Store away from air and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13991-08-7
Molecular Formula C30H24P2
Molecular Weight (g/mol) 446.47
MDL Number MFCD00014081
InChI Key NFRYVRNCDXULEX-UHFFFAOYSA-N
Synonym 1,2-bis diphenylphosphino benzene, dppbz, dppbe, dppben, dppbenz, o-bis diphenylphosphino benzene, 1,2-bis dimethylphosphino benzene, o-phenylenebis diphenylphosphine, phosphine, phenylenebis diphenyl, 1,2-bis diphenylphosphanyl benzene
PubChem CID 498379
IUPAC Name (2-diphenylphosphanylphenyl)-diphenylphosphane
SMILES C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 185°C to 189°C
Quantity 0.25 g
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 446.47
Percent Purity 98%
Chemical Name or Material 1,2-Bis(diphenylphosphino)benzene
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.