Learn More
1,2-Bis(diphenylphosphino)ethane, 97+%
Description
1,2-Bis(diphenylphosphino)ethane is a bidendate ligand used in coordination chemistry to prepare metal complexes. It is involved in the metal-catalyzed allylic alkylation and decarboxylation of allylic esters. It is also used in 1,3-diene synthesis. Further, it is used in cycloaddition reactions and carbonylation reactions. In addition to this, it is used to prepare bis(dicyclohexylphosphino)ethane by hydrogenation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1,2-Bis(diphenylphosphino)ethane is a bidendate ligand used in coordination chemistry to prepare metal complexes. It is involved in the metal-catalyzed allylic alkylation and decarboxylation of allylic esters. It is also used in 1,3-diene synthesis. Further, it is used in cycloaddition reactions and carbonylation reactions. In addition to this, it is used to prepare bis(dicyclohexylphosphino)ethane by hydrogenation.
Solubility
Soluble in acetone, methanol and alcoholic hydrochloric acid.
Notes
Air sensitive. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | 138°C to 143°C |
| Linear Formula | (C6H5)2PCH2CH2P(C6H5)2 |
| Quantity | 25 g |
| Beilstein | 761261 |
| Sensitivity | Air Sensitive |
| Solubility Information | Soluble in acetone,methanol and alcoholic hydrochloric acid. |
| Formula Weight | 398.42 |
| Percent Purity | ≥97% |
| Chemical Name or Material | 1,2-Bis (diphenylphosphino)ethane |
Safety and Handling
| Hazard Category | H315-H319-H335-H500 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 216-769-2 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.