Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Bis(diphenylphosphino)ethane, 97+%

Catalog No. AAA1141922
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1141922 100 g
AAA1141906 5 g
AAA1141914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1141922 Supplier Thermo Scientific Chemicals Supplier No. A1141922
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1663-45-2 | C26H24P2 | 398.43 g/mol

1,2-Bis(diphenylphosphino)ethane is a bidendate ligand used in coordination chemistry to prepare metal complexes. It is involved in the metal-catalyzed allylic alkylation and decarboxylation of allylic esters. It is also used in 1,3-diene synthesis. Further, it is used in cycloaddition reactions and carbonylation reactions. In addition to this, it is used to prepare bis(dicyclohexylphosphino)ethane by hydrogenation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Bis(diphenylphosphino)ethane is a bidendate ligand used in coordination chemistry to prepare metal complexes. It is involved in the metal-catalyzed allylic alkylation and decarboxylation of allylic esters. It is also used in 1,3-diene synthesis. Further, it is used in cycloaddition reactions and carbonylation reactions. In addition to this, it is used to prepare bis(dicyclohexylphosphino)ethane by hydrogenation.

Solubility
Soluble in acetone, methanol and alcoholic hydrochloric acid.

Notes
Air sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1663-45-2
Molecular Formula C26H24P2
Molecular Weight (g/mol) 398.43
MDL Number MFCD00003047
InChI Key QFMZQPDHXULLKC-UHFFFAOYSA-N
Synonym 1,2-bis diphenylphosphino ethane, diphos, ethylenebis diphenylphosphine, bis diphenylphosphine ethane, bis 1,2-diphenylphosphino ethane, phosphine, 1,2-ethanediylbis diphenyl, phosphine, ethylenebis diphenyl, 1,2-bis diphenylphosphine ethane, 1,2-bis diphenylphosphino-ethane, unii-kl33qe52i4
PubChem CID 74267
ChEBI CHEBI:30669
IUPAC Name 2-diphenylphosphanylethyl(diphenyl)phosphane
SMILES C(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 138°C to 143°C
Linear Formula (C6H5)2PCH2CH2P(C6H5)2
Quantity 100 g
Beilstein 761261
Sensitivity Air Sensitive
Solubility Information Soluble in acetone,methanol and alcoholic hydrochloric acid.
Formula Weight 398.42
Percent Purity ≥97%
Chemical Name or Material 1,2-Bis (diphenylphosphino)ethane
Hazard Category H315-H319-H335-H500
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 216-769-2
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.