Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Bis(phenylsulfinyl)ethane Palladium(II) Diacetate 96.0+%, TCI America™
SDP

Catalog No. B32921G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B32921G 1 g
B3292200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B32921G Supplier TCI America Supplier No. B32921G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 858971-43-4
Molecular Formula C18H20O6PdS2
Molecular Weight (g/mol) 502.892
MDL Number MFCD09842752
InChI Key SNNYSJNYZJXIFE-UHFFFAOYSA-L
Synonym White Catalyst
PubChem CID 11562441
IUPAC Name 2-(benzenesulfinyl)ethylsulfinylbenzene;palladium(2+);diacetate
SMILES CC(=O)[O-].CC(=O)[O-].C1=CC=C(C=C1)S(=O)CCS(=O)C2=CC=CC=C2.[Pd+2]

Specifications

Quantity 1 g
Formula Weight 502.89
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Bis(phenylsulfinyl)ethane Palladium(II) Diacetate
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.