Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Bis(trichlorosilyl)ethane 97.0+%, TCI America™
SDP

Catalog No. B129225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B129225G 25 g
B12925G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B129225G Supplier TCI America Supplier No. B129225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2504-64-5
Molecular Formula C2H4Cl6Si2
Molecular Weight (g/mol) 296.924
MDL Number MFCD00013601
InChI Key WDVUXWDZTPZIIE-UHFFFAOYSA-N
PubChem CID 75631
IUPAC Name trichloro(2-trichlorosilylethyl)silane
SMILES C(C[Si](Cl)(Cl)Cl)[Si](Cl)(Cl)Cl

Specifications

Color White
Quantity 25 g
UN Number 2987
Formula Weight 296.92
Percent Purity ≥97.0% (T)
Physical Form Crystalline Lumps
Chemical Name or Material 1,2-Bis(trichlorosilyl)ethane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.