Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2-Bromoethoxy)-2-ethoxybenzene 96.0+%, TCI America™
SDP

Catalog No. B319125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B319125G 25 g
B31915G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B319125G Supplier TCI America Supplier No. B319125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3-8-3259
Molecular Formula C10H13BrO2
Molecular Weight (g/mol) 245.12
MDL Number MFCD02030483
InChI Key IOYHGBZPUZBUTJ-UHFFFAOYSA-N
Synonym 2-2-ethoxyphenoxy ethyl bromide, 1-2-bromoethoxy-2-ethoxybenzene, 2-2-ethoxyphenoxy ethylbromide, 2-2-bromoethoxy phenetole, benzene, 1-2-bromoethoxy-2-ethoxy, 1-2-bromoethoxy-2-ethyoxybenzene, 1-2-bromoethoxy-2-ethoxy-benzene, 2-bromo-1-2-ethoxyphenoxy ethane, tamsulosin impurity i, pubchem13149
PubChem CID 7010183
IUPAC Name 1-(2-bromoethoxy)-2-ethoxybenzene
SMILES CCOC1=CC=CC=C1OCCBr

Specifications

Melting Point 43°C
Color White-Yellow
Quantity 25 g
Formula Weight 245.12
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-(2-Bromoethoxy)-2-ethoxybenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.