Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2-Bromoethoxy)-4-chlorobenzene 98.0+%, TCI America™
SDP

Catalog No. B429825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B429825G 25 g
B42985G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B429825G Supplier TCI America Supplier No. B429825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2033-76-3
Molecular Formula C8H8BrClO
Molecular Weight (g/mol) 235.505
MDL Number MFCD00000617
InChI Key YYFLBDSMQRWARK-UHFFFAOYSA-N
PubChem CID 74854
IUPAC Name 1-(2-bromoethoxy)-4-chlorobenzene
SMILES C1=CC(=CC=C1OCCBr)Cl

Specifications

Melting Point 42°C
Color White
Boiling Point 87°C
Quantity 25 g
Synonym 1-2-bromoethoxy-4-chlorobenzene, benzene, 1-2-bromoethoxy-4-chloro, 2-bromoethyl 4-chlorophenyl ether, 4-chlorophenyl 2-bromoethyl ether, 2-4-chlorophenoxy ethyl bromide, benzene,1-2-bromoethoxy-4-chloro, 2-bromo-1-4-chlorophenoxy ethane, pubchem17076, acmc-1cjap, p-chlorophenoxyethyl bromide
Formula Weight 235.51
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-(2-Bromoethoxy)-4-chlorobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.