Learn More
Sigma Organic Chemistry 1- 2-Bromoethyl -2-meth | 1G | MFCD12197373
Supplier: Sigma Organic Chemistry CBR017031G

1- 2-Bromoethyl -2-meth , 1G
About this Item:
MDL #: MFCD12197373
Molecular Weight: 269.97
UNSPSC Code: 12352200
Chemical Formula: C6H10Br2N2
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.