Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2-Bromoethyl)pyrrole 97.0+%, TCI America™
SDP

Catalog No. B16221G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B16221G 1 g
B16225G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B16221G Supplier TCI America Supplier No. B16221G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 78358-86-8
Molecular Formula C6H8BrN
Molecular Weight (g/mol) 174.041
MDL Number MFCD00191322
InChI Key QBAVHEZVBGASER-UHFFFAOYSA-N
PubChem CID 11651225
IUPAC Name 1-(2-bromoethyl)pyrrole
SMILES C1=CN(C=C1)CCBr

Specifications

Color Yellow
Quantity 1 g
Formula Weight 174.04
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-(2-Bromoethyl)pyrrole
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.